Assay Details
| Gene Symbol: ALDH3B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075518.1 | NP_001068986.1 | 8 - 9 | 1701 | 62 | |
| GenBank mRNA | BC147958.1 | - | 9 - 10 | 1247 | 62 | |
| BT025453.1 | - | 8 - 9 | 1701 | 62 | ||
| BT026328.1 | - | 9 - 10 | 1501 | 62 | ||
More Information
Set Membership: |
|

ALDH3B1
Genomic Map