Assay Details
| Gene Symbol: RTL8C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075522.2 | NP_001068990.1 | 1 | 45 | 66 | |
| GenBank mRNA | BC116150.1 | - | 1 | 48 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC116150.1, NM_001075522.2 transcripts.
Set Membership: |
|

RTL8C
Genomic Map