Assay Details
| Gene Symbol: DHRS7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075545.1 | NP_001069013.1 | 2 - 3 | 290 | 61 | |
| GenBank mRNA | BC114860.1 | - | 2 - 3 | 290 | 61 | |
More Information
Set Membership: |
|

DHRS7C
Genomic Map