Assay Details
| Gene Symbol: SHPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075548.1 | NP_001069016.1 | 5 - 6 | 833 | 58 | |
| GenBank mRNA | BT025410.1 | - | 5 - 6 | 833 | 58 | |
More Information
Set Membership: |
|

SHPK
Genomic Map