Assay Details
| Gene Symbol: TAF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075597.1 | NP_001069065.1 | 3 - 4 | 221 | 133 | |
| GenBank mRNA | BC118136.1 | - | 3 - 4 | 221 | 133 | |
More Information
Set Membership: |
|

TAF13
Genomic Map