Assay Details
Gene Symbol: RHNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075631.1 | NP_001069099.1 | 2 - 3 | 273 | 146 | |
GenBank mRNA | BC116052.1 | - | 2 - 3 | 273 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
