Assay Details
| Gene Symbol: NAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075652.2 | NP_001069120.2 | 5 - 6 | 912 | 84 | |
| GenBank mRNA | BC114778.1 | - | 5 - 6 | 913 | 84 | |
| CK949061.1 | - | 2 - 3 | 224 | 84 | ||
More Information
Set Membership: |
|

NAF1
Genomic Map