Assay Details
| Gene Symbol: IDI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075659.1 | NP_001069127.1 | 2 - 3 | 545 | 60 | |
| GenBank mRNA | BC116133.1 | - | 2 - 3 | 545 | 60 | |
More Information
Set Membership: |
|

IDI1
Genomic Map