Assay Details
| Gene Symbol: DHRS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075687.1 | NP_001069155.1 | 2 - 3 | 415 | 64 | |
| GenBank mRNA | BC118170.1 | - | 2 - 3 | 415 | 64 | |
More Information
Set Membership: |
|

DHRS13
Genomic Map