Assay Details
| Gene Symbol: GPRC5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075741.1 | NP_001069209.1 | 3 - 4 | 1225 | 91 | |
| GenBank mRNA | BC149136.1 | - | 3 - 4 | 1209 | 91 | |
| BT025414.1 | - | 3 - 4 | 1225 | 91 | ||
More Information
Set Membership: |
|

GPRC5B
Genomic Map