Assay Details
| Gene Symbol: DPCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075745.2 | NP_001069213.1 | 4 - 5 | 425 | 69 | |
| GenBank mRNA | BC114010.1 | - | 4 - 5 | 426 | 69 | |
More Information
Set Membership: |
|

DPCD
Genomic Map