Assay Details
Gene Symbol: CMTM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075960.1 | NP_001069428.1 | 2 - 3 | 355 | 70 | |
GenBank mRNA | BC116148.1 | - | 2 - 3 | 355 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
