Assay Details
| Gene Symbol: MRPS31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076015.2 | NP_001069483.1 | 1 - 2 | 171 | 70 | |
| GenBank mRNA | BC118356.1 | - | 1 - 2 | 172 | 70 | |
More Information
Set Membership: |
|

MRPS31
Genomic Map