Assay Details
Gene Symbol: NCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076072.1 | NP_001069540.1 | 2 - 3 | 363 | 120 | |
GenBank mRNA | BC116109.1 | - | 2 - 3 | 363 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
