Assay Details
| Gene Symbol: TSPAN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076119.1 | NP_001069587.1 | 2 - 3 | 498 | 61 | |
| GenBank mRNA | BC118327.1 | - | 2 - 3 | 498 | 61 | |
More Information
Set Membership: |
|

TSPAN5
Genomic Map