Assay Details
| Gene Symbol: CLNS1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076300.1 | NP_001069768.1 | 2 - 3 | 354 | 151 | |
| GenBank mRNA | BC116139.1 | - | 2 - 3 | 354 | 151 | |
More Information
Set Membership: |
|

CLNS1A
Genomic Map