Assay Details
| Gene Symbol: VAMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076302.1 | NP_001069770.1 | 708 | 96 | ||
| GenBank mRNA | BC118341.1 | - | 708 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VAMP7| Gene Symbol: VAMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076302.1 | NP_001069770.1 | 708 | 96 | ||
| GenBank mRNA | BC118341.1 | - | 708 | 96 | ||
Set Membership: |
|