Assay Details
| Gene Symbol: RGS20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076327.1 | NP_001069795.1 | 3 - 4 | 471 | 101 | |
| GenBank mRNA | BC116015.1 | - | 3 - 4 | 471 | 101 | |
More Information
Set Membership: |
|

RGS20
Genomic Map