Assay Details
| Gene Symbol: SLC22A23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076341.1 | NP_001069809.1 | 5 - 6 | 720 | 61 | |
| GenBank mRNA | BC116054.1 | - | 5 - 6 | 720 | 61 | |
More Information
Set Membership: |
|

SLC22A23
Genomic Map