Assay Details
Gene Symbol: PRKAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076358.1 | NP_001069826.1 | 2 - 3 | 305 | 65 | |
GenBank mRNA | BC118242.1 | - | 2 - 3 | 305 | 65 | |
BT030573.1 | - | 2 - 3 | 283 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
