Assay Details
| Gene Symbol: NENF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076419.1 | NP_001069887.1 | 2 - 3 | 239 | 151 | |
| GenBank mRNA | BC116106.1 | - | 2 - 3 | 239 | 151 | |
More Information
Set Membership: |
|

NENF
Genomic Map