Assay Details
| Gene Symbol: EFNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076432.1 | NP_001069900.1 | 2 - 3 | 656 | 59 | |
| GenBank mRNA | BC118089.1 | - | 2 - 3 | 656 | 59 | |
More Information
Set Membership: |
|

EFNA5
Genomic Map