Assay Details
| Gene Symbol: KDELR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076495.1 | NP_001069963.1 | 4 - 5 | 754 | 67 | |
| GenBank mRNA | BC118250.1 | - | 4 - 5 | 754 | 67 | |
More Information
Set Membership: |
|

KDELR1
Genomic Map