Assay Details
| Gene Symbol: SCN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076496.1 | NP_001069964.1 | 2 - 3 | 339 | 73 | |
| GenBank mRNA | BC118258.1 | - | 2 - 3 | 339 | 73 | |
More Information
Set Membership: |
|

SCN1B
Genomic Map