Assay Details
Gene Symbol: CDC5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076542.1 | NP_001070010.1 | 4 - 5 | 543 | 65 | |
GenBank mRNA | BC105417.1 | - | 4 - 5 | 543 | 65 | |
BT025390.1 | - | 4 - 5 | 532 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
