Assay Details
| Gene Symbol: TUBA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077095.1 | NP_001070563.1 | 3 - 4 | 519 | 95 | |
| GenBank mRNA | BC113209.1 | - | 3 - 4 | 519 | 95 | |
More Information
Set Membership: |
|

TUBA8
Genomic Map