Assay Details
| Gene Symbol: PIGW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077109.1 | NP_001070577.1 | 1 - 2 | 89 | 56 | |
| GenBank mRNA | BC116121.1 | - | 1 - 2 | 89 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC116121.1, NM_001077109.1 transcripts.
Set Membership: |
|

PIGW
Genomic Map