Assay Details
| Gene Symbol: ORMDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077133.1 | NP_001070601.1 | 2 - 3 | 315 | 80 | |
| GenBank mRNA | BC114067.1 | - | 2 - 3 | 315 | 80 | |
More Information
Set Membership: |
|

ORMDL1
Genomic Map