Assay Details
| Gene Symbol: DHFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077883.1 | NP_001071351.1 | 2 - 3 | 216 | 100 | |
| GenBank mRNA | BC114162.1 | - | 2 - 3 | 216 | 100 | |
More Information
Set Membership: |
|

DHFR
Genomic Map