Assay Details
| Gene Symbol: BCL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078028.1 | NP_001071496.1 | 2 - 3 | 558 | 138 | |
| GenBank mRNA | BC118326.1 | - | 2 - 3 | 558 | 138 | |
More Information
Set Membership: |
|

BCL10
Genomic Map