Assay Details
| Gene Symbol: TUBA4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078158.1 | NP_001071626.1 | 1 - 2 | 60 | 69 | |
| GenBank mRNA | BC118199.1 | - | 1 - 2 | 60 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118199.1, NM_001078158.1 transcripts.
Set Membership: |
|

TUBA4A
Genomic Map