Assay Details
| Gene Symbol: TIPRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079615.2 | NP_001073083.1 | 3 - 4 | 400 | 62 | |
| GenBank mRNA | BC114024.1 | - | 3 - 4 | 403 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC114024.1, NM_001079615.2 transcripts.
Set Membership: |
|

TIPRL
Genomic Map