Assay Details
Gene Symbol: TIPRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079615.2 | NP_001073083.1 | 5 - 6 | 677 | 147 | |
GenBank mRNA | BC114024.1 | - | 5 - 6 | 680 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
