Assay Details
Gene Symbol: TIPRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079615.2 | NP_001073083.1 | 6 - 7 | 817 | 127 | |
GenBank mRNA | BC114024.1 | - | 6 - 7 | 820 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
