Assay Details
| Gene Symbol: KDELR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079779.1 | NP_001073247.1 | 2 - 3 | 277 | 104 | |
| GenBank mRNA | BC105537.1 | - | 2 - 3 | 277 | 104 | |
More Information
Set Membership: |
|

KDELR2
Genomic Map