Assay Details
| Gene Symbol: PRSS23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080306.1 | NP_001073775.1 | 1 - 2 | 96 | 84 | |
| GenBank mRNA | BC116042.1 | - | 1 - 2 | 96 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC116042.1, NM_001080306.1 transcripts.
Set Membership: |
|

PRSS23
Genomic Map