Assay Details
Gene Symbol: CCDC106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080334.1 | NP_001073803.1 | 2 - 3 | 99 | 120 | |
GenBank mRNA | BC116140.1 | - | 2 - 3 | 99 | 120 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC116140.1, NM_001080334.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
