Assay Details
Gene Symbol: CCDC106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080334.1 | NP_001073803.1 | 3 - 4 | 215 | 73 | |
GenBank mRNA | BC116140.1 | - | 3 - 4 | 215 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
