Assay Details
| Gene Symbol: ANGPTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080345.2 | NP_001073814.1 | 3 - 4 | 799 | 118 | |
| GenBank mRNA | BC105428.1 | - | 3 - 4 | 800 | 118 | |
More Information
Set Membership: |
|

ANGPTL3
Genomic Map