Assay Details
| Gene Symbol: SLAMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174184.4 | NP_776609.2 | 3 - 4 | 782 | 60 | |
| GenBank mRNA | AF329970.1 | - | 3 - 4 | 721 | 60 | |
| BC114833.1 | - | 3 - 4 | 783 | 60 | ||
More Information
Set Membership: |
|

SLAMF1
Genomic Map