Assay Details
| Gene Symbol: UGP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174212.2 | NP_776637.1 | 9 - 10 | 1721 | 69 | |
| GenBank mRNA | BC118181.1 | - | 9 - 10 | 1721 | 69 | |
| L14019.1 | - | 9 - 10 | 1528 | 69 | ||
More Information
Set Membership: |
|

UGP2
Genomic Map