Assay Details
| Gene Symbol: AQP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174702.3 | NP_777127.1 | 1 - 2 | 404 | 65 | |
| GenBank mRNA | AF028005.1 | - | 1 - 2 | 398 | 65 | |
| AH004629.2 | - | 1 - 2 | 420 | 65 | ||
| BC105525.1 | - | 1 - 2 | 404 | 65 | ||
| BT025412.1 | - | 1 - 2 | 430 | 65 | ||
More Information
Set Membership: |
|

AQP1
Genomic Map