Assay Details
Gene Symbol: NDUFA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175826.4 | NP_787020.1 | 2 - 3 | 309 | 59 | |
GenBank mRNA | BC105365.1 | - | 2 - 3 | 310 | 59 | |
X59697.1 | - | 305 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
