Assay Details
| Gene Symbol: SOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201527.2 | NP_963285.2 | 2 - 3 | 292 | 92 | |
| GenBank mRNA | BC105378.1 | - | 2 - 3 | 292 | 92 | |
| BT020988.1 | - | 2 - 3 | 275 | 92 | ||
| L22092.1 | - | 2 - 3 | 250 | 92 | ||
| S67818.1 | - | 2 - 3 | 250 | 92 | ||
More Information
Set Membership: |
|

SOD2
Genomic Map