Assay Details
| Gene Symbol: ARF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015571.3 | NP_001015571.1 | 3 - 4 | 509 | 79 | |
| GenBank mRNA | BC120186.1 | - | 3 - 4 | 511 | 79 | |
| BT020934.1 | - | 2 - 3 | 350 | 79 | ||
More Information
Set Membership: |
|

ARF3
Genomic Map