Assay Details
| Gene Symbol: MYOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040478.2 | NP_001035568.2 | 1 - 2 | 847 | 84 | |
| GenBank mRNA | AB110599.1 | - | 1 - 2 | 633 | 84 | |
| AF093675.1 | - | 1 - 2 | 129 | 84 | ||
| BC120454.1 | - | 1 - 2 | 847 | 84 | ||
More Information
Set Membership: |
|

MYOD1
Genomic Map