Assay Details
| Gene Symbol: MAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040570.2 | NP_001035660.2 | 10 - 11 | 1814 | 77 | |
| GenBank mRNA | BC122604.1 | - | 10 - 11 | 1814 | 77 | |
More Information
Set Membership: |
|

MAG
Genomic Map