Assay Details
| Gene Symbol: FMO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075162.2 | NP_001068630.1 | 7 - 8 | 1302 | 62 | |
| NM_001163274.3 | NP_001156746.2 | 7 - 8 | 1302 | 62 | ||
| GenBank mRNA | BC120448.1 | - | 7 - 8 | 1222 | 62 | |
| CK772691.1 | - | 2 - 3 | 476 | 62 | ||
More Information
Set Membership: |
|

FMO2
Genomic Map