Assay Details
| Gene Symbol: CPTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075182.2 | NP_001068650.1 | 1 - 2 | 65 | 85 | |
| GenBank mRNA | BC120059.1 | - | 1 - 2 | 66 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120059.1, NM_001075182.2 transcripts.
Set Membership: |
|

CPTP
Genomic Map