Assay Details
| Gene Symbol: CPTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075182.2 | NP_001068650.1 | 2 - 3 | 200 | 82 | |
| GenBank mRNA | BC120059.1 | - | 2 - 3 | 201 | 82 | |
More Information
Set Membership: |
|

CPTP
Genomic Map